Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185103.1 | NP_001172032.1 | 9 - 10 | 1129 | 100 | |
XM_021079381.1 | XP_020935040.1 | 1190 | 100 | |||
XM_021079382.1 | XP_020935041.1 | 1190 | 100 | |||
XM_021079383.1 | XP_020935042.1 | 1190 | 100 | |||
XM_021079385.1 | XP_020935044.1 | 1207 | 100 | |||
XM_021079386.1 | XP_020935045.1 | 1207 | 100 | |||
GenBank mRNA | AK237711.1 | - | 9 - 10 | 1166 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |