Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185131.1 | NP_001172060.1 | 6 - 7 | 778 | 85 | |
XM_005663310.3 | XP_005663367.1 | 704 | 85 | |||
XM_021088556.1 | XP_020944215.1 | 837 | 85 | |||
XM_021088557.1 | XP_020944216.1 | 843 | 85 | |||
XM_021088559.1 | XP_020944218.1 | 704 | 85 | |||
GenBank mRNA | AB529527.1 | - | 6 - 7 | 778 | 85 | |
AK236486.1 | - | 7 - 8 | 999 | 85 | ||
AK351226.1 | - | 6 - 7 | 620 | 85 | ||
AK392898.1 | - | 6 - 7 | 705 | 85 | ||
AK398138.1 | - | 6 - 7 | 705 | 85 | ||
AK398787.1 | - | 7 - 8 | 837 | 85 | ||
AY609787.1 | - | 6 - 7 | 1023 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |