Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185179.1 | NP_001172108.1 | 2 - 3 | 157 | 69 | |
NM_001185180.1 | NP_001172109.1 | 1 - 2 | 287 | 69 | ||
XM_005665881.3 | XP_005665938.1 | 267 | 69 | |||
GenBank mRNA | AK347253.1 | - | 2 - 3 | 164 | 69 | |
DY423553.1 | - | 3 - 4 | 327 | 69 | ||
DY427626.1 | - | 1 - 2 | 206 | 69 | ||
EV950782.2 | - | 1 - 2 | 186 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |