Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185185.1 | NP_001172114.1 | 2 - 3 | 339 | 59 | |
GenBank mRNA | AY609909.1 | - | 2 - 3 | 339 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |