Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190157.1 | NP_001177086.1 | 3 - 4 | 317 | 86 | |
XM_005663796.3 | XP_005663853.1 | 423 | 86 | |||
XM_021090958.1 | XP_020946617.1 | 518 | 86 | |||
XM_021090960.1 | XP_020946619.1 | 371 | 86 | |||
GenBank mRNA | AK392706.1 | - | 3 - 4 | 341 | 86 | |
AK400856.1 | - | 4 - 5 | 387 | 86 | ||
AY609522.1 | - | 3 - 4 | 313 | 86 | ||
BX915869.2 | - | 3 - 4 | 336 | 86 | ||
BX919484.2 | - | 4 - 5 | 390 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |