Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190174.1 | NP_001177103.1 | 5 - 6 | 690 | 65 | |
XM_021100259.1 | XP_020955918.1 | 1089 | 65 | |||
XM_021100314.1 | XP_020955973.1 | 877 | 65 | |||
XM_021100355.1 | XP_020956014.1 | 1363 | 65 | |||
GenBank mRNA | AK234741.1 | - | 7 - 8 | 875 | 65 | |
AK395077.1 | - | 7 - 8 | 878 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |