Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190178.1 | NP_001177107.1 | 1 - 2 | 88 | 68 | |
GenBank mRNA | AK239618.1 | - | 1 - 2 | 46 | 68 | |
AK348689.1 | - | 1 - 2 | 42 | 68 | ||
CB479993.1 | - | 4 - 5 | 424 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |