Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190181.1 | NP_001177110.1 | 3 - 4 | 391 | 93 | |
XM_021098029.1 | XP_020953688.1 | 384 | 93 | |||
GenBank mRNA | AK231349.1 | - | 3 - 4 | 310 | 93 | |
AK235777.1 | - | 3 - 4 | 224 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |