Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KATNBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190181.1 | NP_001177110.1 | 7 - 8 | 936 | 105 | |
XM_021098029.1 | XP_020953688.1 | 929 | 105 | |||
GenBank mRNA | AK235777.1 | - | 7 - 8 | 769 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |