Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190196.1 | NP_001177125.1 | 3 - 4 | 505 | 75 | |
GenBank mRNA | AK239025.1 | - | 3 - 4 | 505 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |