Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190210.1 | NP_001177139.1 | 4 - 5 | 679 | 91 | |
XM_005671711.2 | XP_005671768.1 | 812 | 91 | |||
XM_005671712.2 | XP_005671769.1 | 745 | 91 | |||
XM_005671713.3 | XP_005671770.1 | 651 | 91 | |||
XM_005671714.3 | XP_005671771.1 | 1466 | 91 | |||
XM_005671715.2 | XP_005671772.2 | 750 | 91 | |||
XM_013984044.2 | XP_013839498.1 | 775 | 91 | |||
XM_013984045.2 | XP_013839499.1 | 645 | 91 | |||
XM_021074525.1 | XP_020930184.1 | 770 | 91 | |||
GenBank mRNA | AK239144.1 | - | 4 - 5 | 676 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |