Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15H4orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190210.1 | NP_001177139.1 | 6 - 7 | 930 | 83 | |
XM_005671711.2 | XP_005671768.1 | 1063 | 83 | |||
XM_005671712.2 | XP_005671769.1 | 996 | 83 | |||
XM_005671713.3 | XP_005671770.1 | 902 | 83 | |||
XM_005671714.3 | XP_005671771.1 | 1717 | 83 | |||
XM_013984044.2 | XP_013839498.1 | 1026 | 83 | |||
XM_013984045.2 | XP_013839499.1 | 896 | 83 | |||
XM_021074525.1 | XP_020930184.1 | 1021 | 83 | |||
GenBank mRNA | AK239144.1 | - | 6 - 7 | 927 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |