Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190220.1 | NP_001177149.1 | 3 - 4 | 362 | 60 | |
GenBank mRNA | AK234312.1 | - | 3 - 4 | 337 | 60 | |
AK238360.1 | - | 3 - 4 | 362 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |