Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194972.1 | NP_001181901.1 | 9 - 10 | 1112 | 64 | |
XM_021093284.1 | XP_020948943.1 | 4089 | 64 | |||
XM_021093286.1 | XP_020948945.1 | 1310 | 64 | |||
XM_021093287.1 | XP_020948946.1 | 2535 | 64 | |||
GenBank mRNA | GQ329717.1 | - | 9 - 10 | 1112 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |