Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOBOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195116.1 | NP_001182045.1 | 5 - 6 | 1147 | 69 | |
GenBank mRNA | FJ587509.1 | - | 5 - 6 | 1147 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |