Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195343.1 | NP_001182272.1 | 2 - 3 | 486 | 53 | |
GenBank mRNA | GU373693.1 | - | 2 - 3 | 486 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |