Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195359.1 | NP_001182288.1 | 15 - 16 | 1867 | 84 | |
XM_005668865.3 | XP_005668922.1 | 2351 | 84 | |||
XM_005668866.3 | XP_005668923.1 | 2262 | 84 | |||
XM_005668867.3 | XP_005668924.1 | 1787 | 84 | |||
GenBank mRNA | AK237136.1 | - | 1 - 2 | 94 | 84 | |
GU373686.1 | - | 15 - 16 | 1867 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |