Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDR16C5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195368.1 | NP_001182297.1 | 1 - 2 | 332 | 79 | |
XM_013996752.2 | XP_013852206.1 | 611 | 79 | |||
GenBank mRNA | AK344212.1 | - | 2 - 3 | 432 | 79 | |
GU373688.1 | - | 1 - 2 | 332 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |