Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195372.1 | NP_001182301.1 | 4 - 5 | 255 | 94 | |
GenBank mRNA | GU373670.1 | - | 4 - 5 | 255 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Sus scrofa Probe spans exons |