Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199363.1 | NP_001186292.1 | 9 - 10 | 1244 | 82 | |
GenBank mRNA | EU617320.1 | - | 9 - 10 | 1244 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |