Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP8L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204370.1 | NP_001191299.1 | 1 - 2 | 86 | 73 | |
GenBank mRNA | AK239494.1 | - | 1 - 2 | 57 | 73 | |
AK351466.1 | - | 1 - 2 | 57 | 73 | ||
AK392637.1 | - | 1 - 2 | 75 | 73 | ||
AK398852.1 | - | 1 - 2 | 86 | 73 | ||
EU650402.1 | - | 1 - 2 | 62 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |