Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204383.1 | NP_001191312.1 | 2 - 3 | 191 | 79 | |
XM_021091086.1 | XP_020946745.1 | 198 | 79 | |||
GenBank mRNA | AK347593.1 | - | 2 - 3 | 169 | 79 | |
AK349423.1 | - | 2 - 3 | 191 | 79 | ||
AK352228.1 | - | 2 - 3 | 169 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |