Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFN-OMEGA-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166322.1 | NP_001159794.1 | 1 | 347 | 101 | |
GenBank mRNA | GQ415088.1 | - | 1 | 347 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |