Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162890.1 | NP_001156362.1 | 2 | 2042 | 107 | |
XM_013996172.2 | XP_013851626.1 | 2084 | 107 | |||
XM_021085624.1 | XP_020941283.1 | 2310 | 107 | |||
XM_021085625.1 | XP_020941284.1 | 2315 | 107 | |||
GenBank mRNA | AK389264.1 | - | 1 | 1508 | 107 | |
FJ971663.1 | - | 2 | 2042 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Inventoried Both primers and probe map within a single exon |