Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHLDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001174057.1 | NP_001167528.1 | 1 | 247 | 99 | |
GenBank mRNA | GU386348.1 | - | 1 | 247 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |