Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195399.1 | NP_001182328.1 | 1 | 250 | 75 | |
GenBank mRNA | HQ110078.1 | - | 1 | 250 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |