Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139475.1 | NP_001132947.1 | 1 - 2 | 346 | 61 | |
GenBank mRNA | AK238522.1 | - | 2 - 3 | 340 | 61 | |
EU825192.2 | - | 1 - 2 | 346 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |