Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139475.1 | NP_001132947.1 | 4 - 5 | 848 | 79 | |
GenBank mRNA | AK238522.1 | - | 5 - 6 | 842 | 79 | |
EU825192.2 | - | 4 - 5 | 848 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |