Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKMYT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159310.1 | NP_001152782.1 | 3 - 4 | 906 | 73 | |
XM_021085610.1 | XP_020941269.1 | 1260 | 73 | |||
XM_021085611.1 | XP_020941270.1 | 1178 | 73 | |||
XM_021085612.1 | XP_020941271.1 | 1063 | 73 | |||
XM_021085613.1 | XP_020941272.1 | 1306 | 73 | |||
XM_021085614.1 | XP_020941273.1 | 1266 | 73 | |||
XM_021085615.1 | XP_020941274.1 | 1451 | 73 | |||
XM_021085616.1 | XP_020941275.1 | 1184 | 73 | |||
GenBank mRNA | AB495207.1 | - | 3 - 4 | 906 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |