Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161647.1 | NP_001155119.1 | 1 | 1123 | 79 | |
XM_021085620.1 | XP_020941279.1 | 4741 | 79 | |||
XM_021085621.1 | XP_020941280.1 | 4744 | 79 | |||
XM_021085622.1 | XP_020941281.1 | 4123 | 79 | |||
GenBank mRNA | AK349375.1 | - | 1 | 1318 | 79 | |
FJ873107.1 | - | 1 | 1123 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |