Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166045.1 | NP_001159517.1 | 1 - 2 | 836 | 60 | |
GenBank mRNA | GQ478346.1 | - | 1 - 2 | 836 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |