Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167648.1 | NP_001161120.1 | 1 - 2 | 98 | 90 | |
GenBank mRNA | AB529526.1 | - | 1 - 2 | 98 | 90 | |
AK234189.1 | - | 1 - 2 | 237 | 90 | ||
AK392968.1 | - | 1 - 2 | 201 | 90 | ||
AK395009.1 | - | 1 - 2 | 182 | 90 | ||
AK400157.1 | - | 1 - 2 | 195 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |