Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167651.1 | NP_001161123.1 | 4 - 5 | 652 | 79 | |
XM_013979136.2 | XP_013834590.1 | 2287 | 79 | |||
GenBank mRNA | AB529861.1 | - | 4 - 5 | 652 | 79 | |
AK231437.1 | - | 4 - 5 | 717 | 79 |
Gene Symbol: LOC110255553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK349852.1 | - | 3 - 4 | 525 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Sus scrofa Probe spans exons |