Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190254.1 | NP_001177183.1 | 1 - 2 | 432 | 75 | |
XM_005665895.3 | XP_005665952.1 | 3805 | 75 | |||
XM_021097998.1 | XP_020953657.1 | 1805 | 75 | |||
GenBank mRNA | AK396692.1 | - | 3 - 4 | 764 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |