Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190254.1 | NP_001177183.1 | 3 - 4 | 685 | 80 | |
XM_005665895.3 | XP_005665952.1 | 4058 | 80 | |||
XM_021097998.1 | XP_020953657.1 | 2058 | 80 | |||
GenBank mRNA | AK350071.1 | - | 2 - 3 | 258 | 80 | |
AK396692.1 | - | 5 - 6 | 1017 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |