Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190280.1 | NP_001177209.1 | 7 - 8 | 1881 | 89 | |
XM_021078467.1 | XP_020934126.1 | 2270 | 89 | |||
GenBank mRNA | HM046519.1 | - | 7 - 8 | 1881 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |