Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195356.1 | NP_001182285.1 | 3 - 4 | 578 | 73 | |
GenBank mRNA | AK240357.1 | - | 4 - 5 | 639 | 73 | |
GU373666.1 | - | 3 - 4 | 578 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |