Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195356.1 | NP_001182285.1 | 4 - 5 | 745 | 69 | |
GenBank mRNA | AK240357.1 | - | 5 - 6 | 806 | 69 | |
GU373666.1 | - | 4 - 5 | 745 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |