Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206352.1 | NP_001193281.1 | 1 - 2 | 146 | 83 | |
GenBank mRNA | AK234315.1 | - | 1 - 2 | 146 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |