Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244863.1 | NP_001231792.1 | 6 - 7 | 722 | 83 | |
XM_013985196.2 | XP_013840650.1 | 1054 | 83 | |||
XM_021077266.1 | XP_020932925.1 | 1719 | 83 | |||
XM_021077267.1 | XP_020932926.1 | 999 | 83 | |||
GenBank mRNA | AK349737.1 | - | 6 - 7 | 724 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |