Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KANSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244880.1 | NP_001231809.1 | 7 - 8 | 1023 | 85 | |
XM_005664181.3 | XP_005664238.1 | 85 | ||||
XM_005664182.3 | XP_005664239.1 | 85 | ||||
XM_013997991.2 | XP_013853445.1 | 85 | ||||
XM_013997992.2 | XP_013853446.1 | 85 | ||||
XM_021091033.1 | XP_020946692.1 | 85 | ||||
XM_021091034.1 | XP_020946693.1 | 85 | ||||
GenBank mRNA | AK234157.1 | - | 7 - 8 | 1030 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |