Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244893.1 | NP_001231822.1 | 3 - 4 | 801 | 95 | |
GenBank mRNA | AK236177.1 | - | 3 - 4 | 765 | 95 | |
AK237169.1 | - | 3 - 4 | 801 | 95 | ||
AK395565.1 | - | 3 - 4 | 801 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |