Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244899.1 | NP_001231828.1 | 3 - 4 | 650 | 62 | |
XM_021072055.1 | XP_020927714.1 | 668 | 62 | |||
XM_021072056.1 | XP_020927715.1 | 669 | 62 | |||
GenBank mRNA | AK346287.1 | - | 3 - 4 | 650 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |