Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244918.1 | NP_001231847.1 | 11 - 12 | 1318 | 109 | |
XM_005666324.3 | XP_005666381.1 | 1467 | 109 | |||
GenBank mRNA | AK349556.1 | - | 11 - 12 | 1318 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |