Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244932.1 | NP_001231861.1 | 3 - 4 | 537 | 82 | |
XM_013981911.2 | XP_013837365.2 | 373 | 82 | |||
GenBank mRNA | AK351541.1 | - | 3 - 4 | 537 | 82 | |
AK389451.1 | - | 3 - 4 | 545 | 82 | ||
AK395786.1 | - | 3 - 4 | 541 | 82 | ||
AK400781.1 | - | 3 - 4 | 537 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |