Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTRF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244934.1 | NP_001231863.1 | 6 - 7 | 966 | 114 | |
XM_013992474.2 | XP_013847928.1 | 843 | 114 | |||
XM_021072417.1 | XP_020928076.1 | 860 | 114 | |||
GenBank mRNA | AK231749.1 | - | 6 - 7 | 685 | 114 | |
AY609949.1 | - | 6 - 7 | 966 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |