Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244943.1 | NP_001231872.1 | 2 - 3 | 380 | 71 | |
XM_005665240.3 | XP_005665297.2 | 430 | 71 | |||
GenBank mRNA | AK349783.1 | - | 2 - 3 | 380 | 71 | |
AK393463.1 | - | 2 - 3 | 399 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |