Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244943.1 | NP_001231872.1 | 4 - 5 | 637 | 116 | |
XM_005665240.3 | XP_005665297.2 | 687 | 116 | |||
GenBank mRNA | AK349783.1 | - | 4 - 5 | 637 | 116 | |
AK393463.1 | - | 4 - 5 | 656 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |