Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244969.1 | NP_001231898.1 | 2 - 3 | 598 | 103 | |
XM_021079468.1 | XP_020935127.1 | 644 | 103 | |||
GenBank mRNA | AK235534.1 | - | 2 - 3 | 598 | 103 | |
AK346598.1 | - | 2 - 3 | 598 | 103 | ||
AK391076.1 | - | 2 - 3 | 619 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |