Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244971.2 | NP_001231900.2 | 3 - 4 | 2154 | 92 | |
XM_005653406.3 | XP_005653463.1 | 1261 | 92 | |||
XM_005653407.3 | XP_005653464.1 | 357 | 92 | |||
XM_021093325.1 | XP_020948984.1 | 2100 | 92 | |||
GenBank mRNA | AK395469.1 | - | 3 - 4 | 753 | 92 | |
AY610046.1 | - | 3 - 4 | 463 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |